Qualatex® XC 5C Powder-Free Latex Fingercots, QRP
Supplier: PIP
5CS 5CM 5CL 5CXL
14003-425CS 1363.35 CAD
14003-425 14003-427 14003-429 14003-431
Qualatex® XC 5C Powder-Free Latex Fingercots, QRP
Finger Cots Latex Finger Cots
Eliminates contamination risks from silicone oils, skin salts, particulates, and plasticizers.
  • Packaged for ISO 5 (Class 100) applications.
  • Eliminates contamination risks from silicone oils, skin salts, particulates, and plasticizers
  • Unique, Stand-Up Pouch- easy product identification
The 5C Qualatex® powder-free cleanroom latex finger cots are packaged for ISO 5 (Class 100) applications
Applications: Discrete components, hard drive assemblies, die bonding, wire attach, disk drives/thin film heads, qoptics, finishing, cleaning & handling of precision instruments.
Compliant with ASTM and IEST-RP-CC 005.3 standards. Eliminates contamination risks from silicone oils, skin salts, particulates, and plasticizers. Finger cots are packaged in gas-impremeable film flushed with semiconductor grade N2. Unique, Stand-Up Pouch provides easy product identification.
Certifications: Compliant with ASTM and IEST-RP-CC 005.3 standards.
Caution: This product contains natural rubber latex which may cause allergic reactions.