Qualatherm® 59G Dry Thermal Protection Gloves, 14", QRP
Supplier: QRP
Qualatherm®
59GXL 59GL
89041-062CS 527.04 CAD
89041-062 32889-019
Qualatherm® 59G Dry Thermal Protection Gloves, 14", QRP
Gloves
QRP 59G Qualatherm gloves are RoHS compliant gloves, packaged for ISO 5 Class 100 cleanroom applications.
  • Dry contact temperatures from –210 to 1000 °F (–134 to 537 °C) with no charring, ash, or residue
  • No PCB, asbestos, or fiberglass
Applications: Diffusion furnaces, quartz wafer boards, liquid gas flask handling (ultra cold), hot/dry glassware handling and metal fabrication.
The middle layer is 100% virgin wool knit with a high terry pile for excellent insulation. Completely nylon-lined to minimize particulate shed during donning and removal.