Qualatherm® 50G Heat and Cold-Resistant 14" Gloves, QRP
Supplier: PIP
50GL 50GXL
32885-931PK 750.29 CAD
32885-931 89041-060
Qualatherm® 50G Heat and Cold-Resistant 14" Gloves, QRP
Gloves
The QRP 50G gloves are RoHS compliant, ambidextrous protection gloves designed for use in DRY heat/cold handling environments.
  • Packaged for ISO 5 (Class 100)
  • Withstands dry excursion temperatures from -210°F to 1400°F (-134°C to 650°C), with no charring, ash or residue
  • ESD protection Static Dissipative 10⁹
  • No PCB, asbestos, or fiberglass
The knit outer shell withstands dry excursion temperatures from -210°F to 1400°F (-134°C to 650°C), with no charring, ash or residue.
Not designed for use in moist, steamy, or wet environments. Completely nylon-lined to minimize linting and particulate generation during donning and removal, when the opportunity is greatest to generate lint and particles. The middle layer is 100% virgin wool knit with a high terry pile for excellent insulation. These gloves are cleanroom 100 compatible.
Applications: Diffusion furnaces, quartz wafer boats, liquid gas flask handling (ultra cold), hot/dry glassware handling and metal fabrication.
Certifications: ESD Safe to ANSI/ESD S20.20-2014.