Qualatex® 2C Low Powder Latex Finger Cots, QRP
Supplier: PIP
2CL 2CS 2CM 2CXL
56613-911CS 2429.71 CAD
56613-911 56613-977 56613-988 56613-999
Qualatex® 2C Low Powder Latex Finger Cots, QRP
Finger Cots Latex Finger Cots
QRP 2C Finger Cots provide natural latex protection from skin salts, flakes, and oils, dyes, chemical, and solvent splash.
  • For general , personal, and product protection
  • Sheer film for maximum dexterity
  • Economical
  • For industrial use only
High level of comfort, abrasion resistance, and tactile sensitivity. Textured, rolled, nominal 2.75" length. 3 mil, tissue weight, sheer film for maximum dexterity and relief from finger fatigue.
Caution: This product contains natural rubber latex which may cause allergic reactions.