PolyTuff® 28G PolyTuff Conductive Gloves, QRP
Supplier: PIP
28GXL 28GS 28GM 28GL
89041-058CS 4014.72 CAD
89041-058 32889-020 32889-022 32889-024
PolyTuff® 28G PolyTuff Conductive Gloves, QRP
Gloves
Tough, sheer PolyTuff polyurethane (1.5 mil) film affords a high degree of tactile sensitivity.
  • Packaged for ISO 5 (Class 100) applications
  • Static dissipative 10⁶ ohms per square surface unit
  • ESD- safe for Class I and II devices
  • Tough solvent process polyurethane, sulfur-free - no carbon shedding
  • Ergonomic Comfort Curve™ hand specific design
Applications: Wafer fabrication, printed circuit boards, IC handling, disk drives, electronics industries; automotive painting, electrostatic painting; communications systems, cellular device fabrication; militaryMore Product Information