PolyTuff® 27G PolyTuff Conductive Gloves, QRP
Supplier: PIP
27GXL 27GS 27GL
89041-056CS 5033.86 CAD
89041-056 32891-130 32891-146
PolyTuff® 27G PolyTuff Conductive Gloves, QRP
Gloves
Tough, sheer PolyTuff polyurethane (1.5 mil) film affords a high degree of tactile sensitivity.
  • Packaged for ISO (Class 100) applications
  • Static dissipative 10⁶ ohms per square surface unit
  • ESD safe for Class I and II devices
  • Tough solvent process polyurethane, sulfur-free - no carbon shedding
  • Ergonomic Comfort Curve™ hand specific design
Applications: Wafer fabrication, printed circuit boards, IC handling, disk drives, electronics industries; automotive painting, electrostatic painting; communications systems, cellular device fabrication; military avionics, sensors, robotics fabrication.
Tough, sheer PolyTuff polyurethane (1.5 mil) film affords a high degree of tactile sensitivity coupled with outstanding resistance to snags, punctures, and abrasions. Seamless paired RH/LH gloves and Comfort Curve™ design minimize hand and finger fatigue. No carbon shedding. Resistant to many common solvents, including TCE and IPA, per ASTM F-739.
Certifications: Compliant with MIL-STD-1686D, conductive per MIL-B-81705. ISO 5 (class 100) processed and packaged in double-bagged, static-safe packaging. ESD safe for Class 0, I and II devices.