Orion™ Star™ A323 Dissolved Oxygen Portable Meter, Thermo Scientific
Supplier: Thermo Fisher Scientific
STARA3230 STARA3235
89206-356EA 1869.9 CAD
89206-356 89206-358
Orion™ Star™ A323 Dissolved Oxygen Portable Meter, Thermo Scientific
Electrochemistry Meters Oxygen Meters
Accurately monitor dissolved oxygen (optical/polarographic) and temperature with the Orion™ Star™ A323 RDO/DO Portable Meter for challenging field testing.
  • Informative, easy-to-read backlit graphic display with clear onscreen instructions simplifies operation
  • Capture critical readings with measurement stability indicator and selectable read modes
  • Auto-Read mode holds a stable reading, continuous mode shows changing readings with hold option, and Timed mode logs data at specific intervals
  • Data log collects and date/time stamps up to 5,000 measurement sets
More Product Information