Orion™ Star™ A123 Dissolved Oxygen Portable Meter, Thermo Scientific
Supplier: Thermo Fisher Scientific
STARA1230 STARA1235 STARA1236
89206-298EA 1110.71 CAD
89206-298 89206-300 76076-750
Orion™ Star™ A123 Dissolved Oxygen Portable Meter, Thermo Scientific
Electrochemistry Meters Oxygen Meters
Take simple dissolved oxygen and temperature measurements with the Orion™ Star™ A123 DO Portable Meter with easy operation for basic field testing.
  • Measure dissolved oxygen in percent saturation or mg/L with temperature using polarographic DO probes
  • Calibrate using water-saturated air or custom value (Winkler titration)
  • Automatically compensate DO readings using ATC temperature, manual salinity input, and manual barometric pressure
  • Simplified, easy-to-read display shows main measurement with temperature, stability indicator, and electrode condition icon
More Product Information