Orion™ Star™ A213 Dissolved Oxygen Benchtop Meter, Thermo Scientific
Supplier: Thermo Fisher Scientific
STARA2130 STARA2135 STARA2136
89206-320EA 2955 CAD
89206-320 89206-322 89206-324
Orion™ Star™ A213 Dissolved Oxygen Benchtop Meter, Thermo Scientific
Electrochemistry Meters Oxygen Meters
Reliably measure dissolved oxygen (RDO optical / polarographic) and temperature with the Orion™ Star™ A213 RDO/DO Benchtop Meter for advanced laboratory analysis.
  • Automatically compensate dissolved oxygen readings using ATC temperature, manual salinity input, and built-in barometer for barometric pressure readings
  • Informative, easy-to-read backlit graphic display with clear onscreen instructions simplifies operation
  • Capture critical readings with measurement stability indicator and selectable read modes
  • Auto-Read mode holds a stable reading, continuous
More Product Information