Orion™ Versa Star Pro™ DO Benchtop Meter, Thermo Scientific
Supplier: Thermo Fisher Scientific
VSTAR30 VSTAR32
89206-386EA 4187 CAD
89206-386 89206-388
Orion™ Versa Star Pro™ DO Benchtop Meter, Thermo Scientific
Electrochemistry Meters Oxygen Meters
Get premium performance and flexibility with the Orion™ Versa Star Pro™ Meter with DO/RDO Module for dissolved oxygen (optical/polarographic) and temperature.
  • Onscreen measurement stability indicator lets users know exactly when measurements are ready
  • Auto-read mode holds a stable reading, continuous mode shows changing readings, Timed mode logs data at specific intervals, and Single-Shot mode holds a reading after a preset time is reached
  • Audible alarms automatically notify users when a set offset value, high/low limit value, and calibration time interval are reached
  • Calibration log stores ten most recent calibrations per parameter
  • Ensure measurement integrity by viewing the active calibration log while taking readings
  • Three-year meter and module warranty
Get high accuracy, premium performance, and optimal flexibility with the Orion™ Versa Star Pro™ benchtop meter with dissolved oxygen/RDO module.
Orion™ Versa Star Pro™ benchtop meters are designed to meet the most challenging application with ease. Delivering rich functionality and features, users can optimally configure the meter system to meet changing requirements. These meters offer interchangeable measurement modules that allow multiple users to customize four separate channels to meet their specific requirements. Easily view measurements on a stunning color display with adjustable text sizes. Take the guesswork out of meter calibration and setup with onscreen instructions available in a variety of local languages. Enjoy quick and simple data entry using the numeric keypad. Easily access, update, and switch preferred measurement settings, using ten password-protected methods per channel. Uniformly mix solutions using up to two meter-controlled stirrer probes that are easy to position and can be quickly rinsed between samples. Measure dissolved oxygen in percent saturation or mg/L with temperature using Orion RDO optical DO probes or Orion polarographic DO probes and automatic sensor recognition. Automatically compensate dissolved oxygen readings using ATC temperature, manual salinity input, and built-in barometer for barometric pressure readings. Customize measurements to show calibration history or simplify to show an extra large dissolved oxygen reading. Non-volatile meter memory preserves data and settings, even in the event of power loss. IP54 rated housing resists dust and splashes, and can be wall-mounted.
Meet the most challenging applications for dissolved oxygen and temperature. Easily view measurements on the stunning color display and customize the four meter channels with interchangeable modules. Use an RDO optical or polarographic DO probe and calibrate using water-saturated air, air-saturated water, custom value (Winkler), or zero point. Log up to 2000 data point sets with time/date stamp and transfer to a printer or computer.
Certifications: CE, TUV 3-1, FCC Class A.
Ordering information: For compatible DO probe, see Orion™ item 083005MD. For compatible buffer solutions, see Orion™ item 80513.