innuPREP Virus DNA/RNA Kit, Analytik Jena
Supplier: Analytik Jena CA
845-KS-4800050 845-KS-4800250
CA10755-384EA 1402.27 CAD
CA10755-384 CA10755-386
innuPREP Virus DNA/RNA Kit, Analytik Jena
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The innuPREP Virus DNA/RNA allows researchers to purify viral ssDNA or dsDNA and ssRNA simultaneously from serum, plasma or other cell-free bodily fluids, from tissue, paraffin or swab samples, or from cell cultures.
  • Simultaneous isolation of viral DNA and RNA from a variety of starting materials
  • Patented DC technology: rapid lysis and efficient binding
  • Extraction method based on the use of spin filters
  • Optimum removal of inhibitors ensures trouble-free use of nucleic acids in subsequent applications
  • Recommended for samples with unknown viruses
  • Includes carrier mix with internal DNA and RNA extraction control
The innuPREP Virus DNA/RNA allows researchers to purify viral ssDNA or dsDNA and ssRNA simultaneously from serum, plasma or other cell-free bodily fluids, from tissue, paraffin or swab samples, or from cell cultures. A novel extraction chemistry known as “Dual Chemistry” (DC) technology guarantees ability to isolate highly pure viral nucleic acids of excellent quality. The use of a spin filter membrane maximizes DNA and RNA yields. In addition, having a number of different extraction protocols makes it possible to adapt the innuPREP Virus DNA/RNA Kit to the starting material used. One major advantage of this kit is the time saved by isolating nucleic acids simultaneously, particularly when using starting materials in which the viral contamination is not clear.
Specifications
Starting material: Serum, plasma, cell-free bodily fluids, supernatants from cell cultures (400 µl each); Tissues and biopsies of up to 20 mg; Swab samples
Quality: Positive PCR and TaqMan® real-time PCR testing results
Extraction time: Approx. 25 minutes