AlbumiNZ™ Bovine Protease-Reduced BSA (from Serum)
Supplier: MP Biomedicals
AlbumiNZ
0219989890 0219989891 0219989850 0219989825 0219989880
10273-300EA 5484.24 CAD
10273-300 10273-278 10273-298 10273-296 IC219989825
AlbumiNZ™ Bovine Protease-Reduced BSA (from Serum)
Proteins and Peptides
AlbumiNZ™ Protease-reduced BSA is useful for immunoassays where proteases may degrade proteins or where peroxidase or alkaline phospahatase may hydrolyze substrates.
  • No Protease activity detected
  • Endotoxin ≤ 1 EU/mg
  • No Mycoplasma detected
  • No virus detected
  • Moisture: 1.1% (KF)
  • Proteases: ≤ 0.0002 units/mg
AlbumiNZ™ Protease-reduced BSA is chromatographically purified from New Zealand-sourced bovine plasma in an ISO quality system assuring complete traceability and consistent high quality. There is no detectable enzymatic activity.
Certain conformational and primary-sequence epitopes of BSA are suspected allergens in human beef and milk allergies.
Protease Reduced