CottonWipe I Wipers, CleanTex™, Advantus
Supplier: Advantus
CT304A CT306 CT312
33344-002PK 208.84 CAD
33344-002 33344-004 33344-012
CottonWipe I Wipers, CleanTex™, Advantus
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Cotton Wipe I is a strong, double sided twill pattern cotton wiper woven in a cross section of 118×60 threads per square inch.
  • 100% cotton
  • Bias cut and tight-weave construction
  • Autoclavable
  • Extremely durable, highly absorbent, and heat resistant
  • Low-lint surface and edges that will not unravel minimize product contamination
  • Excellent resistance to buildup of electrostatic charge
For use with cleaning, polishing, and burnishing of metallic and nonmetallic magnetic media disk surfaces. Cleaning diffusion furnace equipment in water fabrication and other equipment in areas where high temperatures prevent the use of synthetic wipers. Removing aqueous and organic solvent spills.
The product is manufactured with long-staple cotton yarn to eliminate free-floating fibers on fabric surface.