innuPREP Plant RNA Kit, Analytik Jena
Supplier: Analytik Jena CA
845-KS-2060050 845-KS-2060250
CA10755-392EA 1602.09 CAD
CA10755-392 CA10755-798
innuPREP Plant RNA Kit, Analytik Jena
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The chemistry underlying the innuPREP Plant RNA Kit, which has been specially adapted for isolating plant materials (e.g. leaves, caulis, root, blossom), guarantees highly efficient lysis and effectively deactivates endogenous and exogenous RNases.
  • Ready-to-use total plant RNA in just 30 minutes after homogenization
  • Selection of two integrated lysis buffer systems for optimum adaptation to an extremely wide variety of plant species and components
  • Selective prefiltration step to prevent DNase digestion
  • Optimized extraction chemistry eliminates the need for toxic β-mercaptoethanol
The chemistry underlying the innuPREP Plant RNA Kit, which has been specially adapted for isolating plant materials (e.g. leaves, caulis, root, blossom), guarantees highly efficient lysis and effectively deactivates endogenous and exogenous RNases. The innuPREP Plant RNA Kit contains two lysis buffers in order to process as wide a range as possible of different plant materials, particularly plant components with widely differing characteristics. Specifically, the Lysis Buffer RL is a universal buffer and the Lysis Buffer PL has been specifically adapted to difficult plants such as roses or potatoes. The total extracted RNA is of excellent quality can be used directly in subsequent applications.
Specifications
Starting material: Various types of plant samples (100mg max.); Fresh or frozen plant material
Column binding capacity: approx. 100 µg RNA
Average purity: 1.8 – 2.0
Extraction time: Approx. 30 minutes after homogenization
Average yield: Up to 70µg, depending on the type and quantity of the sample