innuPREP Plant DNA Kit, Analytik Jena
Supplier: Analytik Jena CA
845-KS-1060050 845-KS-1060250
CA10755-592EA 1028.6 CAD
CA10755-592 CA10755-594
innuPREP Plant DNA Kit, Analytik Jena
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The innuPREP Plant DNA Kit has been specially developed for quickly and easily isolating DNA from an extremely wide variety of plant starting materials (such as leaves, stems, roots, flowers, etc).
  • Isolation of highly pure DNA free of plant inhibitors and secondary metabolic products
  • Suitable for use with an extremely wide variety of plant materials
  • High yields from up to 100mg starting material
  • 3-lysis buffer system for optimized lysis of plant material
  • Specific guidelines according to sample type
  • Tested for fresh and frozen samples (including dry archived material), leaves, wood, seeds, needles, and fruits
The innuPREP Plant DNA Kit has been specially developed for quickly and easily isolating DNA from an extremely wide variety of plant starting materials (such as leaves, stems, roots, flowers, etc). In addition to efficient sample digestion, the extraction routine also includes a prefiltration step to effectively minimize unlysed plant components. The DNA is then bound to a spin filter column using a novel binding buffer, after which it is washed and then eluted in a separate elution tube. The extracted nucleic acid is then immediately available for a number of downstream applications and can be stored for future applications without any trouble.
Specifications
Starting material: Various plant materials (max. 100 mg); Fresh, frozen or dried plant material
Column binding capacity: >50µg DNA
Average purity: 1.7 – 2.0
Extraction time: Approx. 30 – 40 minutes
Average yield: Approx. 3 – 25µg, depending on the type and starting quantity of the plant material