Lamps for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
G1314-60100 2140-0590 G1103-60001 2140-0813 5182-1530 110715400 2140-0820 5190-0917 G1971-60025
CA76508-244EA 1249.41 CAD
CA76508-244 CA76514-584 CA76515-174 CA2140-0813 CA5182-1530 CAAG110715400 76537-440 76537-442 CAAGG197160025
Lamps for HPLC, Agilent Technologies
Lights Deuterium Lamps
A wide selection of certified, long-life lamps for Agilent HPLC systems.
  • Contain RFID technology for more convenience and ease of use
  • Tested for noise and drift specifications, correct operating voltage, light intensity, and proper alignment
  • Improved coating process increases lamp lifetimes
  • Agilent deuterium lamps are designed with a much narrower aperture, providing increased light intensity and decreased noise for an appreciably higher signal-to-noise ratio
  • Higher sensitivity extends detection capabilities and improves qualification at trace levels, for
More Product Information