BS-480 Chemistry Analyzer, without ISE Module, MedTest DX

Supplier: MedTest DX
BS-480UPS BS-480
10023-808EA 637740.76 CAD
10023-808 10023-812
BS-480 Chemistry Analyzer, without ISE Module, MedTest DX
Clinical Diagnostic Supplies Clinical Analysers
The MedTest 480 Ensemble provides exceptional value and convenience by bringing together everything your mid-volume laboratory needs to perform clinical chemistry testing

  • Robust hardware
  • Enhanced liquid system
  • Economy usage
  • Easy to perform maintenance
  • Intelligent probe system
  • Advanced software
  • User-friendly interface
  • Real-time QC status monitoring
  • Traceable test results

MedTest will manage and oversee every step of your customized project from installation and operator training to testing of your new analyzer, reagents, calibrators/standards, quality control materials, and consumables.

More specifications
Throughput: 500 tests/ hr. with ISE and 400 tests/ hr. without ISE
Measuring Principle: Absorbance Photometry, Turbidimetry
Sample Tray: 90 positions for primary or secondary tubes and sample cups
Sample Volume: 1.5 µL, step by 0.1 µL
Sample Probe: liquid level detection, clot detection, horizontal and vertical collision protection
Sample & Reagent Barcode Reading Includes: Codabar, ITF, Code 128, Code 39, UPC/EAN, Code 93; Bi-directional LIS interface transmission
Reagent Tray: 80 positions
Light Source: Halogen-tungsten lamp
Photometer: Reversed, optics, grating photometry
Wavelengths (nm): 340, 380, 412, 450, 505, 546, 570, 605, 660, 700, 740, 800
Absorbance Range: 0-3.3 Abs
Resolutions: 0.0001 Abs
Operating System: Windows 8, Windows 10
Interface: RS-232, network port, USB/ parallel port
Power Supply: 110-130 V, 60 Hz, 1500 VA
Dimensions (W x D x H in.): 46 x 28 x 45
Weight: 661 lbs

Ordering information: The BS-480 UPS accessory must be purchased; it is recommended for consistent power being supplied to the BS-480 instrument. Analyzer includes the instrument, computer, touchscreen monitor, and keyboard. The analyzer has a 12 month warranty and additional years of service are available for purchase. The following services are provided with purchase: installation, operator training on-site, telephone technical support, remote diagnostics if the lab has internet access, on-site engineer if needed and chemistry and drugs of abuse screening reagents. For the analyzer with the ISE (electrolyte) module, contact your VWR representative.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR