Orion™ Star™ A329 pH/ISE/Conductivity/Dissolved Oxygen Portable Multiparameter Meter, Thermo Scientific
Supplier: Thermo Fisher Scientific
STARA3290 STARA3295
89206-372EA 3163.95 CAD
89206-372 89206-374
Orion™ Star™ A329 pH/ISE/Conductivity/Dissolved Oxygen Portable Multiparameter Meter, Thermo Scientific
Electrochemistry Meters pH and Multi-Parameter Meters
Accurately measure pH, ion concentration, conductivity, dissolved oxygen and temperature with the Orion™ Star™ A329 pH/ISE/Conductivity/RDO/DO Portable Meter.
  • Combined convenience with the benefits of a pH/ISE meter, conductivity meter and dissolved oxygen meter in one easy-to-use system
  • Perform up to a five point conductivity calibration
  • Auto-ranging conductivity values offer consistent readings across entire measurement range without need to recalibrate
  • Use linear, non-linear, EP (USP) or off conductivity temperature compensation with 5, 10, 15, 20 or 25°
More Product Information