VWR® Advanced Dry Block Heaters, 230 V (Export Only)

Supplier: VWR International
75838-266 75838-274 75838-276 75838-288 75838-290 75838-300 75838-302 75838-320 75838-322 75838-324
75838-266EA 3316.56 CAD
75838-266 75838-274 75838-276 75838-288 75838-290 75838-300 75838-302 75838-320 75838-322 75838-324
VWR® Advanced Dry Block Heaters, 230 V (Export Only)
Block Heaters Digital Block Heaters
Designed for applications that require repeatable results and superior temperature stability.

Multi-purpose units are ideal for incubation and activation of cultures, enzyme reactions, immunoassays, melting/boiling points, and a variety of other laboratory procedures

230 V model, not for use in USA
New design with smaller footprint
Exceptional temperature uniformity and stability
Heated lid model reduces condensation in sample lids
LED display for temperature and time
Heats to 120 °C (245 °F)
Holds interchangeable, modular heating blocks
5 year warranty on parts and labor
Available with a NISTMore Product Information
Order Now

SPECIFICATIONS

Environmentally Preferable Waste Reducing Waste Reducing Waste Reducing Waste Reducing Waste Reducing Waste Reducing
Temperature range 5° above ambient to 120°C (248°F) 5° above ambient to 120°C (248°F) 5° above ambient to 120°C (248°F) 5° above ambient to 120°C (248°F) 5° above ambient to 120°C (248°F) 5° above ambient to 120°C (248°F)
Temperature stability ±0.2°C ±0.2°C ±0.2°C ±0.2°C ±0.2°C ±0.2°C
Temperature uniformity ±0.2°C (within the Block @ 37°C) ±0.2°C (within the Block @ 37°C) ±0.2°C (within the Block @ 37°C) ±0.2°C (within the Block @ 37°C) ±0.2°C (within the Block @ 37°C) ±0.2°C (within the Block @ 37°C)
Environmental certification RoHS RoHS RoHS RoHS RoHS RoHS

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR