Human Recombinant CNTF (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
100-59AF-500UG 100-59AF-5UG 100-59AF-20UG 100-59AF-100UG 100-59AF-1MG
10821-620EA 4067.38 CAD
10821-620 10821-632 10821-634 10821-636 10821-638
Human Recombinant CNTF (from E. coli)
Proteins and Peptides
Ciliary neurotrophic factor (CNTF) is a neurotrophic factor that promotes the survival of neuronal cell populations, neurite outgrowth, and neurotransmitter synthesis. CNTF also plays an important protective role during nervous system injury.
  • High quality
  • Low endotoxin
  • Affordable