Human Recombinant RELM-beta (Animal Free) (from E. coli)
Supplier: Fujifilm Irvine Scientific
100-160AF-5UG 100-160AF-25UG 100-160AF-1MG
10821-696EA 146.47 CAD
10821-696 10821-698 10821-700 10821-702 10822-348
Human Recombinant RELM-beta (Animal Free) (from E. coli)
Proteins and Peptides
Resistin-like molecule-beta (RELM-β) is a member of the RELM family of secreted proteins containing conserved C-terminus cysteines. The RELM family consists of Resistin (FIZZ3), RELM-α (FIZZ1), RELM-β (FIZZ2), and RELM-ɣ (FIZZ4). Resistin and RELM-β are the only RELM family members found in humans, whereas all four RELM family members are present in rodents. RELM-β functions to increase fibroblast proliferation and differentiation, resulting in airway remodelling and increased inflammation.
  • High Quality
  • Low Endotoxin
  • Affordable
Caution: For research use only.