Human Recombinant GDNF (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
100-02AF-10UG 100-02AF-100UG 100-02AF-1MG 100-02AF-500UG
10821-672EA 159.6 CAD
10821-672 10821-674 10821-676 10821-678 10822-012
Human Recombinant GDNF (from E. coli)
Proteins and Peptides
Glial cell-derived neurotrophic factor (GDNF) is a neurotrophic factor that signals through a multicomponent receptor system to activate receptor tyrosine kinase RET signaling. GDNF promotes dopamine uptake, prevents motor neuron apoptosis, and supports the survival and differentiation of neurons.
  • High quality
  • Low endotoxin
  • Affordable