SeaPlaque® Agarose
Supplier: LONZA WALKERSVILLE INC. CA
50100 50101 50110 50111
CA12001-898EA 1357.55 CAD
CA12001-898 CA12001-900 CA12001-902 CA12001-904
SeaPlaque® Agarose
Media for Microbiology Dehydrated Media for Microbiology
SeaPlaque® is a low-melting, temperature agarose perfect for DNA/RNA recovery, In-Gel PCR/Ligation/Transformation, tissue culture and viral plaque assays.
  • Quality tested to certify performance
  • DNAse/RNAse free
  • Low melting temperature
SeaPlaque® is the original low melting temperature agarose
SeaPlaque® GTG® is a low melting temperature agarose designed for the separation of DNA fragments between 100 bp to 23,000 bp and is ideally suited for direct enzymatic manipulation of nucleic acids in remelted agarose. SeaPlaque® GTG® Agarose is tested and certified for ligation-transformation and restriction digestion using remelted agarose protocols. SeaPlaque™ GTG agarose has no detectable DNase or RNase activity.
This molecular biology grade agarose produces gels with greater sieving properties and higher clarity than standard melting temperature agarose. The low melting temperature of SeaPlaque™ agarose makes it ideal for preparative DNA and RNA electrophoresis, while its low gelling temperature is ideal for cloning of tissue culture cells and viral plaque assays.