BioClean-D™ Disposable Sleeve Covers, Ansell
Supplier: Ansell Canada
S-BDSC-L BDSC-L S-BDSC-L BDSC-L
75834-078CS 1012.06 CAD
75834-078 75972-646 CA75834-078 CA75972-646
BioClean-D™ Disposable Sleeve Covers, Ansell
Disposable Garments
BioClean-D™ disposable sleeve covers provides comfort and protection.
  • Extra-long 500 mm length
  • Exceptional antistatic properties
  • Low-linting
  • Available in sterile and non sterile formats
  • Durable CleanTough™ material construction
  • Tested against permeation standard ASTM D6978-05
  • ISO Class 4 compatible
Offering exceptional comfort and protection, single-use are constructed from antistatic, low-linting CleanTough™ material. They feature elasticated upper arms and cuffs. This 'one size fits' all cover has been tailored for quick and simple donning.
Certifications: Manufactured in a facility holding ISO 9001:2008 and 89/686/EEC Article 11b certifications.
Conforms to Category 3 - Complex Design Personal Protective Equipment PPE Directive 89/686/EEC.
Tested against standard EN ISO 13034 protective clothing against liquid chemicals (Type PB[6]).
Tested against standard EN ISO 14325 protective clothing against chemicals.
Tested against standard EN 1149-1 protective clothing - electrostatic properties.
Tested against permeation standard ASTM D6978-05 for cytotoxic drugs.
Processed in an NEBB certified ISO Class 4 environment.
Sterilized in accordance with ISO 11137-1:2006.
Packaging: Sleeves are packaged one pair per sealed inner PE bag; 15 inner bags per sealed outer PE bag; six outer bags per lined carton (90 pairs).