BioClean-C™ Non sterile Chemotherapy Protective Apron with Sleeves, Ansell
Supplier: Ansell Canada
BCAS-L BCAS-S BCAS-XL BCAS-M BCAS-L BCAS-S BCAS-XL BCAS-M
75834-004CS 1214.09 CAD
75834-004 75834-080 75834-082 75834-124 CA75834-004 CA75834-080 CA75834-082 CA75834-124
BioClean-C™ Non sterile Chemotherapy Protective Apron with Sleeves, Ansell
Disposable Garments
BioClean-C™ Non sterile chemotherapy protective apron with sleeves offers comfort and protection.
  • Tested against ASTM D6978-05 using a minimum of nine cytotoxic drugs
  • Ultrasonically bonded seams with protective tape
  • Tie tapes at rear
  • Adjustable neck fastening
  • 100% polyester elasticated cuffs
  • Lightweight, low-linting, and hard wearing CleanTough™ material
  • ISO Class 4 compatible
It provides maximum protection against a range of cytotoxic drugs. Manufactured from lightweight, low-linting, and hard wearing CleanTough™ material. The apron features an adjustable neck fastening, with tie tapes at the rear, ultrasonically bonded sleeve seams with protective tape, and 100% polyester elasticated cuffs.
Certifications: Manufactured in a facility holding ISO 9001:2008 and 89/686/EEC Article 11b certifications.
Conforms to Category 3 - Complex Design Personal Protective Equipment PPE Directive 89/686/EEC.
Tested against standard EN ISO 13034 protective clothing against liquid chemicals (Type PB [6]).
Tested against standard EN 14325 protective clothing against chemicals.
Tested against permeation standard ASTM D6978-05 for cytotoxic drugs.
Processed in an NEBB certified ISO Class 4 environment.
Packaging: Aprons are packaged one piece per sealed inner PE bag; one inner bag per sealed outer PE bag; 40 outer bags per lined carton (40 pieces).