BioClean™ Clearview™ Autoclavable Silicone Rubber Goggles, Ansell
Supplier: Ansell Healthcare
BCAH
75833-999PK 5366.14 CAD
75833-999
BioClean™ Clearview™ Autoclavable Silicone Rubber Goggles, Ansell
Goggles
BioClean™ Clearview™ Autoclavable Silicone Rubber Goggles are optically correct for clear vision.
  • Super-soft silicone rubber frame
  • Indirect ventilation system
  • Toughened polycarbonate, anti-scratch, optically correct lens
  • Adjustable hook and eye clip for easier donning
  • Non-linting, latex-free head band
  • Tested to withstand >40 autoclave cycles of 30 minutes duration at 121 °C (250 °F)
  • Can be worn over eyeglasses
  • ISO Class 5 compatible
Constructed in super-soft, lightweight silicone rubber to provide unrivalled comfort, BioClean™ Clearview™ Autoclavable Cleanroom Goggles feature an indirect ventilation system, toughened anti-scratch polycarbonate lens, and can be worn over eyeglasses.
Goggles feature a silicone rubber frame, toughened polycarbonate lens, and latex-free silicone head band with polypropylene hooks.
Provided nonsterile.
Certifications: Manufactured in a facility holding ISO 9001:2008.
Processed in an NEBB certified ISO Class 4 cleanroom environment.
Packaging: Goggles are packaged one piece per sealed PE bag (to be removed prior to autoclaving); 12 bags per inner box; five boxes per lined carton (60 pieces).