Market Forge Sterilmatic® STM-ED/EDX, Steam Pressure Digital Sterilizer, Crown Steam
Supplier: Market Forge
STM-ED STM-EDX
10127-644EA 37540.8 CAD
10127-644 10127-646
Market Forge Sterilmatic® STM-ED/EDX, Steam Pressure Digital Sterilizer, Crown Steam
Sterilizers Autoclaves (Steam Sterilizers)
The sterilizing cycle is fully automatic with time, temperature and venting controlled by the digital microprocessor controller. The sterilizing temperature can be set in a range from 225 °F (107 °C) and 250 °F (121°C).
  • LCD display touch screen
  • Temperature in °F or °C and pressure in PSI or kPa
  • Sterilizing temperatures from 225 to 250°F (107 to 121 °C)
  • Choose from FAST or SLOW exhaust
  • Built-in USB port and RS232 serial port for optional printer
  • Programable controller to store multiple temp/time/vent settings
  • Maintains set temperature with built-in
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Temperature range 107...121 °C (225...250 °F)
Operating pressure 1 atm, 100 kPa (15 psi)