BioClean™ Extra™ Sterile Latex Gloves, Ansell
Supplier: Ansell Canada
BLAS-L BLAS-M BLAS-S BLAS-XL BLAS-L BLAS-M BLAS-S BLAS-XL
75834-088CS 1460.81 CAD
75834-088 75834-090 75834-092 75834-094 CA75834-088 CA75834-090 CA75834-092 CA75834-094
BioClean™ Extra™ Sterile Latex Gloves, Ansell
Gloves
BioClean™ Extra™ sterile latex gloves offer unbeatable comfort and are elbow length for extra protection.
  • Elbow length protection
  • Flexible and comfortable
  • Textured for improved grip
  • Double-donnable
  • Non-particulating EasyTear™ packaging
With a textured surface for enhanced grip, this natural-colored glove is ambidextrous and features a beaded cuff for stability on the arm.
Certifications: Manufactured in a facility holding ISO 9001: 2008 and 89/686/EEC Article 11b certifications.
Conforms to Category 3 - Complex Design Personal Protective Equipment PPE Directive 89/686/EEC.
Processed in an NEBB certified ISO Class 4 environment.
Complies with the requirements of Glove Standard EN420:2003 and EN374-1, 2 and 3:2003.
Physical properties comply with European Medical Glove Standard EN455-2:2009.
Sterilized in accordance with ISO 11137-1:2006.
Packaging: Gloves are packaged one pair per inner PE wallet; one wallet per sealed EasyTear™ PE pouch; 10 pouches per sealed outer PE bag; 20 outer bags per lined carton (200 pairs).