BioClean™ Nerva™ Non sterile Nitrile Gloves, Ansell
Supplier: Ansell Canada
BNAL-L BNAL-M BNAL-S BNAL-XL BNAL-XXL BNAL-L BNAL-M BNAL-S BNAL-XL BNAL-XXL
75834-036CS 1405.87 CAD
75834-036 75834-038 75834-040 75834-042 75834-044 CA75834-036 CA75834-038 CA75834-040 CA75834-042 CA75834-044
BioClean™ Nerva™ Non sterile Nitrile Gloves, Ansell
Gloves
BioClean™ Nerva™ non-sterile nitrile gloves offer excellent comfort and elbow length protection.
  • Elbow length protection
  • Textured for improved grip
  • Non-particulating EasyTear™ packaging
With excellent ESD properties and latex-free construction, these ambidextrous gloves feature improved double-donning properties, enhanced grip, and a beaded cuff for added strength. Available in a blue or white finish.
Certifications: Manufactured in a facility holding ISO 9001: 2008.
Processed in an NEBB certified ISO Class 4 environment.
Complies with the requirements of European Glove Standards EN 420:2003 and EN 374-1, 2 & 3:2003.
Physical properties comply with European Medical Glove Standard EN 455-2:2009.
Packaging: Gloves are packaged 100 pieces per sealed inner PE bag; one inner PE bag per sealed outer PE bag; 10 outer bags per lined carton (1000 pieces).These parts are set up as Case of 500 = 500 pairs = 1000 pieces