Shimpo LCD Contact Handheld Tachometers, Nidec Shimpo America
Supplier: Nidec Shimpo America
DT-105A-S12 DT-105A
75836-548EA 838.96 CAD
75836-548 75836-630
Shimpo LCD Contact Handheld Tachometers, Nidec Shimpo America
Tachometers
The Series DT-105A Tachometer with microprocessor technology has heralded a superior standard in tachometer accuracy and capabilities for measuring and recording speeds of rotational, linear and surface plus total length.
  • Outstanding Accuracy allows use in the strictest applications
  • Sophisticaed laser beam detection up to 20 feet provides operator safety
  • Exceptionally rugged die-cast aluminum housing
  • 10 Measurement memory capacity allows quick retrieval of stored data
  • Long lasting battery life up to 40 hours
  • Rear Threaded inserts provides option to permanently mount the device.
  • CE Mark and NIST Certification included provide guarantee of quality performance
Offering unprecedented confidence, the DT-105A battery-operated tachometers feature the same rugged, high quality components that have made them the benchmark in the industry. These user-friendly, value-packed units feature memory storage; quick adjustment unit’s knob; length functions; retrieval of last, maximum and minimum values; plus each unit comes standard with a free N.I.S.T. certificate. Units are available with an LCD or vibrant red LED display. Incorporating the precise technology and user-friendly functions as the DT-105A, the DT-105A-S12 provides an exceptional accuracy in measuring speed and length via its 12” wheel. The larger wheel rotates more slowly than a standard 6” wheel, enabling reliable and safe registration of high speeds.
Certifications: CE, NIST.
Ordering information: Supplied with NIST Certificate 2 Cone adapters, funnel adapter, 3.5" extension shaft, carrying case, master 6" or 12" circumference wheel, 2AA batteries and contact adapter.