QuEChERS Performance Standard B, Restek
Supplier: RESTEK CORP
31154
75834-628EA 147.82 CAD
75834-628
QuEChERS Performance Standard B, Restek
Standards Organic Standards
Designed for use in all QuEChERS methods for pesticides in fruits and vegetables, including the original unbuffered method, AOAC 2007.01, and EN15662.
  • Ideal for initial method evaluations and ongoing method performance validations.
  • Optimized blend of chemically compatible analytes for maximum stability and shelf life.
  • Precise formulation improves data quality and operational efficiency; spend more time running samples and less time sourcing and preparing standards
  • Quantitatively analyzed to confirm the composition and stability of each mixture
  • Produced and tested in accordance with ISO Guide 34 and 17025 accreditation.
It has 7 components.
Certifications: QuEChERS Performance Standards.