8270 Matrix Spike Mix, Restek
Supplier: RESTEK CORP
31687 31687-15 33073
75834-742EA 286.77 CAD
75834-742 75834-744 75835-896
8270 Matrix Spike Mix, Restek
Standards Organic Standards
Ordering information: *1,2-diphenylhydrazine (8270-listed analyte) decomposes to azobenzene (mix component) in the injector.
**N-nitrosodiphenylamine (8270-listed analyte) decomposes to diphenylamine (mix component) in the injector.