EPA 524.3 Reference Standards (5 separate mixes, including a MegaMix standard), Restek
Supplier: RESTEK CORP
30014 30015 30016 30013 30017
75834-382EA 90.06 CAD
75834-382 75834-384 75834-386 75834-390 75835-532
EPA 524.3 Reference Standards (5 separate mixes, including a MegaMix standard), Restek
Standards Environmental Analysis Standards
In support of the U.S. Safe Drinking Water Act (SDWA), Restek offers a complete set of EPA 524.3 reference standards for the monitoring of purgeable organic compounds in drinking water—using as few as three ampuls! In addition, this collection of certified reference materials (CRMs) also covers the seven volatile organic compounds (VOCs) included in the Unregulated Contaminant Monitoring Rule 3 (UCMR3), which requires monitoring of all public drinking water systems with 10,000 or more customers.
  • Full 82-component EPA 524.3 list using as few as three ampuls—reduce prep time and chances for error or contamination
  • EPA 524.3 VOA MegaMix ampul includes oxygenates group—no need to order separately
  • Volatile gases prepared separately—replace shorter-life components without wasting money on full list
  • Two options for internal and surrogate standards—separate or combined mix
  • Certified reference materials (CRMs) manufactured and QC-tested in Restek’s ISO-accredited labs—satisfy your ISO requirements
  • Also ideal for surface water and groundwater testing.