VWR® Standard Dry Block Heaters, 230 V (Export Only)

Supplier: VWR International
75838-268 75838-272 75838-292 75838-304
75838-268EA 857.93 CAD
75838-268 75838-272 75838-292 75838-304
VWR® Standard Dry Block Heaters, 230 V (Export Only)
Block Heaters Analogue Block Heaters
New designed high wattage, constant temperature VWR® standard dry block heaters with smaller footprint are economical, versatile and compact.

  • 230 V model, not for use in USA
  • Exceptional temperature uniformity
  • Holds interchangeable modular heating blocks
  • Heats to 150 °C (302 °F)
  • 5-Year-Warranty on parts & labor
  • Manufactured in the USA

Multi-purpose units are ideal for incubation and activation of cultures, enzyme reactions, immunoassays, melting/boiling points, and a variety of other laboratory procedures

Each model accepts separate interchangeable modular blocks, which accommodate various sample enclosures such as vials, microplates, PCR strips, and a wide variety of tubes. Each block contains a thermometer well, and features close tube-and-block contact for maximum heat retention.

All units offer two separate temperature adjustment knobs providing reproducible low temperature and high temperature control. Low range knob adjusts from ambient to 100 °C (212 °F) and high range knob adjusts from 75 to 150 °C (167 to 302 °F).

Operating Conditions: Units can be run in conditions from 18 to 33 °C (64 to 91 °F) 20% to 80% relative humidity, non-condensing.

Certifications: CE Marked, TUV listed, RoHS Compliant.

Accessories information: Heaters require a VWR® modular heating block for operation (sold separately). Over 40 modular heating block options are available.

Ordering information: Unit must be ordered with heating block(s). All units include a 234cm (92") detachable, Euro and UK style plug, and a Euro to Swiss adapter.

Caution: To avoid possible electrical hazard, do not fill well or block opening with water or other fluids. Unit is designed as a dry bath/incubator. 230 V model, not for use in USA. Corresponding model with voltage suitable for use in US is also available, please see separate product page.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR