PCB-Free Transformer Oil, Restek
Supplier: RESTEK CORP
32424 32425
75834-972EA 47.4 CAD
75834-972 75834-974
PCB-Free Transformer Oil, Restek
Standards Petroleum Industry Standards
ASTM Method D4059-00 is used for determining PCB concentrations in various types of transformer oil using GC-ECD detection
The analyst must dilute transformer oil samples in a solvent prior to injection. The oil in the sample has been shown to quench the ECD. Calibration mixtures of PCBs in transformer oil must be prepared and diluted identically to eliminate the detector quenching bias resulting when samples are analyzed.
Certifications: ASTM Method D4059-00 (PCB Standards in Oil)