Diesel Fuel #2 (ASTM Class 5 Accelerant), Restek
Supplier: RESTEK CORP
31233 31234 31235 31236
75834-654EA 73.91 CAD
75834-654 75834-656 75835-622 75835-624
Diesel Fuel #2 (ASTM Class 5 Accelerant), Restek
Standards Petroleum Industry Standards
Certified reference material (CRM)
These solutions are prepared from a single-source (one-refinery) product. The weathered materials indicate the percent weight loss from the original material.
Certifications: Weathered Petrochemical Standards