Polywax Standards, Restek
Supplier: RESTEK CORP
36225 36227
10058-072EA 47.57 CAD
10058-072 75835-440
Polywax Standards, Restek
Standards Petroleum Industry Standards
These high molecular weight hydrocarbon waxes are useful for simulated distillation and other high-temperature GC work.