Maintenance Sorbents, 3M Company
Supplier: 3M
7000029594 7000126020
CA56616-320CS 146.23 CAD
CA56616-320 CA56616-391 CA56616-395 CA56616-398
Maintenance Sorbents, 3M Company
Spill Control Materials
Maintenance sorbent mini-booms can be placed around the base of equipment to help contain fluids that may leak or spill.
  • For use on most non-aggressive liquids
  • Available in three lengths
  • Ideal for placing around equipment
  • Helps prevent leaks/spills from spreading to adjacent areas
  • Scrim on the outside of mini-boom identifies the use while containing particulate Sorbent material
Maintenance sorbent mini-booms can be placed around the base of equipment to help contain fluids that may leak or spill, preventing spread to adjacent work areas, One gallonMore Product Information