Cryo.s™ Biobanking Tubes with Datamatrix, Greiner Bio-One
Supplier: Greiner Bio-One
976561 977561 978561
75832-302CS 1831.13 CAD
75832-302 75832-304 75832-306
Cryo.s™ Biobanking Tubes with Datamatrix, Greiner Bio-One
Vials
Cryo.s™ biobanking tubes represent ideal solutions for the space-efficient storage of biological samples in large-scale biorepositories.
  • Stackable
  • Compact footprint
  • Screw caps on tubes
  • Rotation stoppers on rack bottom
  • Unique 2D codes
  • Tubes have no graduation or writing area
These racks with 96 Cryo.s™ biobanking sample storage tubes are pre-produced with Datamatrix barcoding on tubes and rack. Tubes and caps are made of durable polypropylene, and racks can hold a total capacity of 96 Cryo.s™ biobanking tubes.
Ordering information: Includes manual decapping tool.