Partial Immersion Thermometers, Non Mercury, Chemglass
Supplier: Chemglass
CG-3503-11 CG-3503-12
80072-180EA 87.71 CAD
80072-180 80072-182
Partial Immersion Thermometers, Non Mercury, Chemglass
Thermometers Analog Thermometers
Thermometer is mineral spirit filled to eliminate workplace and environmental hazards associated with mercury.
All ranges have 1° subdivisions. Thermometer has a white back and a 6 to 7 mm outer diameter, and is calibrated for 76 mm (3") immersion.