Student Thermometer
Supplier: United Scientific Supplies
THPCF1 THPC06
470223-768EA 12.8 CAD
470223-768 470349-778
Student Thermometer
Thermometers Analog Thermometers
No-roll triangle shape.
  • Partial immersion
  • White backing
  • Features re-usable plastic storage tube
This student grade thermometer is filled with kerosene with red dye.