Ezwaste™ XL Solvent Waste System Replacement Chemical Exhaust Filters, Foxx Life Sciences
Supplier: Foxx Life Sciences
330-0909-OEM 330-0910-OEM
75805-400EA 825.41 CAD
75805-400 75805-402
Ezwaste™ XL Solvent Waste System Replacement Chemical Exhaust Filters, Foxx Life Sciences
Filters Liquid Process Filters
Replacement Chemical Exhaust Filters for the Ezwaste™ XL Solvent Waste System are designed to be an affordable solution for HPLC liquid waste disposal.
  • Each system is manufactured to safely collect waste from two HPLC machines
EZwaste™ XL Solvent Waste filter replacements are supplied in a pack of 1.
Depending on application, carbon exhaust filter life cycle may vary.
This system features an activated carbon filter which traps volatile organic compounds from being released into the air. This prevents chemicals from being inhaled by lab workers and provides an easy mechanism of disposal for vapor wastes. The system also features a closed loop design which protects against leaks and spills, improving overall lab safety.