TSX Series High-Performance −20 °C Manual Defrost Enzyme Freezers, Thermo Scientific
Supplier: Thermo Fisher Scientific
TSX2320ED TSX2320EA TSX3020EA TSX3020ED
75839-108EA 21060.75 CAD
75839-108 75839-116 75839-180 76320-310 76320-312 76320-314
TSX Series High-Performance −20 °C Manual Defrost Enzyme Freezers, Thermo Scientific
Freezers Laboratory / General Purpose Freezers
Thermo Scientific™ TSX Series high-performance -20°C manual defrost enzyme freezers offer a sustainable choice for enzyme storage.
  • Whisper quiet with a sound level of 52 dBA
  • V-drive technology
  • Up to 50% less energy usage than conventional refrigerant models
  • Lower HVAC costs as a result of less heat emission into the environment
  • Easy to use microprocessor control with setpoint security and alarms
  • Compatible with wireless monitoring systems and remote alarm contacts
  • Standard door locks provide comfort and assurance
  • Ergonomic door handle