TSX Series High-Performance Pharmacy Refrigerators, Automatic Defrost, Thermo Scientific
Supplier: Thermo Fisher Scientific
TSX2305PA TSX2305PD TSX5005PA TSX5005PD TSX3005PA TSX3005PD TSX1205PA TSX1205PD
75839-044EA 18463.83 CAD
75839-044 75839-104 75839-112 76208-594 76208-596 76320-218 76320-220 76320-222 76320-270 76320-272 76320-296
TSX Series High-Performance Pharmacy Refrigerators, Automatic Defrost, Thermo Scientific
Refrigerators
TSX Series High-Performance Pharmacy Refrigerators are the quiet, sustainable choice for medical and pharmacy-grade refrigeration.
  • Whisper quiet with a sound level of 52 dBA
  • V-drive technology and Smart View compatibility
  • Up to 40% less energy usage than conventional refrigerant models
  • Convenient, sliding drawer baskets for simplified inventory and access
  • Lower HVAC costs as a result of less heat emission into the environment
  • Easy to use microprocessor control with interior light, setpoint security and alarms
  • Compatible with wireless monitoring systems and remote alarm contacts
  • Standard door locks provide comfort and assurance
  • Ergonomic door handle
Thermo Scientific™ TSX Series high-performance pharmacy refrigerators are designed with features that support sample protection and sustainability objectives for the storage of pharmaceuticals, vaccines, chemotherapy and other medical and pharmacy-grade storage requiring 2° to 8°C
Designed to safely store pharmaceuticals, vaccines, chemotherapy and other medical-grade storage requiring a temperature range of 3°C to 7°C. Units are whisper quiet with a sound level of 52 dBA, enabling end users to work alongside the refrigerator without distraction. Unique V-drive technology is designed to detect usage patterns such as door openings when a higher compressor speed is needed and periods of stability where the compressor runs at a lower speed, saving energy without compromising protection. Refrigerators offer up to 40% less energy usage than conventional refrigerant models.
Refrigerators feature an automatic defrost and a microprocessor display.
Operating temperature range of 2° to 8°C, factory setpoint at 5°C.
Certifications: Refrigerators use natural, R290 refrigerants, making the TSX Series compliant with the U.S. Environmental Protection Agency's Significant New Alternatives Policy (SNAP). Manufactured in an award-winning, zero-waste to landfill facility. Refrigerators are UL, cUL certified.
Accessories information: Optional chart recorders available.