TSX Series High-Performance Blood Bank Refrigerators, Automatic Defrost, Thermo Scientific
Supplier: Thermo Fisher Scientific
TSX2304BA TSX2304BD TSX5004BA TSX5004BD TSX1204BA TSX1204BD TSX3004BD TSX3004BA
75839-040EA 25280 CAD
75839-040 75839-092 75839-100 76208-598 76208-600 76320-226 76320-228 76320-230 76320-232 76320-308 76320-316
TSX Series High-Performance Blood Bank Refrigerators, Automatic Defrost, Thermo Scientific
Refrigerators
Thermo Scientific™ TSX Series high-performance blood bank refrigerators offer a sustainable choice for whole blood and blood components refrigeration.
  • Stores up to 385 blood bags (450 ml whole blood)
  • Whisper quiet with a sound level of 52 dBA
  • V-drive technology and Smart View compatibility
  • Up to 40% less energy usage than conventional refrigerant
  • Convenient, fully extendable and adjustable stainless steel drawers
  • Compatible with wireless monitoring systems and remote alarm contacts
  • Standard door locks provide comfort and assurance
  • Ergonomic door handle
  • Standard factory-installed chart recorder
Thermo Scientific™ TSX Series high-performance blood bank refrigerators are designed with features that support sample protection and sustainability objectives for the storage of whole blood and blood components
Units are whisper quiet with a sound level of 52 dBA, enabling end users to work alongside the refrigerator without distraction. Unique V-drive technology is designed to detect usage patterns such as door openings when a higher compressor speed is needed and periods of stability where the compressor runs at a lower speed, saving energy without compromising protection. These refrigerators offer up to 40% less energy usage than conventional refrigerant models.
Refrigerators feature an automatic defrost and a microprocessor display.
Operating temperature range of 2 to 8 °C, factory setpoint at 4 °C.
Lower HVAC costs as a result of less heat emission into the environment. Easy to use microprocessor control with interior light, setpoint security and alarms.
Certifications: Designed to meet the strict requirements established by the AABB and FDA for the storage of whole blood and blood components. Class I MDD, Class II for FDA. Refrigerator uses natural, R290 refrigerants, making the TSX Series compliant with the U.S. Environmental Protection Agency's Significant New Alternatives Policy (SNAP). Manufactured in an award-winning, zero-waste to landfill facility. All refrigerators are UL, cUL, FDA certified. Refrigerator with European plug is CE, UL, cUL, FDA certified.