TSX Series High-Performance Lab Refrigerators, Automatic Defrost, Thermo Scientific

Supplier: Thermo Fisher Scientific
TSX2305GA TSX2305SA TSX2305GD TSX2305SD TSX5005SA TSX5005SD TSX5005GA TSX5005GD TSX4505GA TSX4505GD TSX3005SA TSX3005SD TSX3005GA TSX3005GD TSX1205SA TSX1205SD TSX1205GA TSX1205GD
75839-042EA 16274 CAD
75839-042 75839-046 75839-094 75839-102 75839-106 75839-114 76208-586 76208-588 76208-590 76208-592 76208-610 76208-612 76320-212 76320-214 76320-216 76320-256 76320-258 76320-260 76320-262 76320-264 76320-266 76320-268 76320-292 76320-294
TSX Series High-Performance Lab Refrigerators, Automatic Defrost, Thermo Scientific
Refrigerators
Thermo Scientific™ TSX Series high-performance laboratory refrigerators offer a sustainable choice for clinical and laboratory refrigeration.

  • Designed to safely store medical and laboratory storage
  • Whisper quiet with a sound level of 52 dBA
  • V-drive technology
  • Up to 40% less energy usage than conventional refrigerant models
  • Choice of glass or solid door models
  • Lower HVAC costs as a result of less heat emission into the environment
  • Easy to use microprocessor control with interior light, setpoint security and alarms
  • Compatible with wireless monitoring systems and remote alarm contacts
  • Standard door locks provide comfort and assurance
  • Ergonomic door handle

Thermo Scientific™ TSX Series high-performance lab refrigerators are designed with features that support sample protection and sustainability objectives for the storage of media, reagents, chemotherapy, pharmaceutical and other medical and laboratory-grade storage requiring 3 to 7 °C

Designed to safely store medical and laboratory storage, including vaccines, pharmaceuticals, media and reagents requiring a temperature range of 2°C to 8°C. Units are whisper quiet with a sound level of 52 dBA, enabling end users to work alongside the refrigerator without distraction. Unique V-drive technology is designed to detect usage patterns such as door openings when a higher compressor speed is needed and periods of stability where the compressor runs at a lower speed, saving energy without compromising protection. These refrigerators offer up to 40% less energy usage than conventional refrigerant models.

Refrigerators feature an automatic defrost and a microprocessor display.

Manufactured in an award-winning, zero-waste to landfill facility. Operating temperature range of 2 to 8 °C, factory setpoint at 5 °C.

5 year warranty on parts and labor, 10 year warranty on compressor parts.

Certifications: All refrigerators are UL, cUL certified. Refrigerators with Europran plug are CE, UL, cUL certified. Refrigerators use natural, R290 refrigerants, making the TSX Series compliant with the U.S. Environmental Protection Agency's Significant New Alternatives Policy (SNAP).

Accessories information: Optional chart recorders available.

Caution: Note: Dimensions shown below are W×D×H.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR