Glass Plates, Square, Chemglass
Supplier: Chemglass
CG-1904-07 CG-1904-17 CG-1904-22 CG-1904-27 CG-1904-31 CG-1904-32 CG-1904-41 CG-1904-42 CG-1904-01 CG-1904-02 CG-1904-06 CG-1904-11 CG-1904-12 CG-1904-16 CG-1904-18 CG-1904-21 CG-1904-36 CG-1904-37 CG-1904-38 CG-1904-39 CG-1904-40
80061-602EA 31.91 CAD
80061-602 80061-622 80061-632 80061-642 80061-650 80061-652 80061-670 80061-672 80061-590 80061-592 80061-600 80061-610 80061-612 80061-620 80061-624 80061-630 80061-660 80061-662 80061-664 80061-666 80061-668
Glass Plates, Square, Chemglass
Electrophoresis Plates and Sheets
Made of Pyrex® or Duran® borosilicate glass.
The 1/16,1/8 and 1/4" thick plates have edges that are cut and swiped. The 1/2" thick plates have ground edges.