SP Wilmad-LabGlass Stainless Steel McCarter Reaction Clamps, SP Industries
Supplier: SP Industries
LG-7316-106 LG-7316-108
14008-880EA 183.45 CAD
14008-880 14008-882 14008-884 14008-886 14008-888
SP Wilmad-LabGlass Stainless Steel McCarter Reaction Clamps, SP Industries
Clamps General Purpose Clamps
These stainless steel horseshoe-shaped clamps are uniquely designed to connect two pieces of glass with flat flanges of the same diameter
Adjustable tension springs secure even pressure over the flange area, and may be slid on or off easily. This facilitates assembly and disassembly of apparatus setups.