SP Wilmad-LabGlass Gay-Lussac Adjusted Specific Gravity Bottles, SP Industries
Supplier: SP Industries
LG-3540-104 LG-3540-100 LG-3540-102 LG-3540-110 LG-3540-108 LG-3540-106
14203-350EA 665.06 CAD
14203-350 14203-144 14210-304 14209-678 14207-798 14203-352
SP Wilmad-LabGlass Gay-Lussac Adjusted Specific Gravity Bottles, SP Industries
Specific Gravity Bottles
These borosilicate glass specific gravity bottles are accurately adjusted at 20°C to determine the density of liquid samples.
While the joints have Standard Taper dimensions, the capillary stoppers are not interchangeable since the final adjustment for volume is made on the stopper. Each bottle and its stopper carry the same identifying number.