Lovibond® MD 100 COD for Photometer, Tintometer
Supplier: Tintometer
276120
10752-412EA 1586.26 CAD
10752-412
Lovibond® MD 100 COD for Photometer, Tintometer
Environmental Test Kits
The MD 100 COD Photometer provides photometric detection to measure the COD concentration from 0 to 15000 mg/l O2 and a simple button layout, robust waterproof design and large backlit display make the MD100 easy to use and ideal for testing in the field, laboratory or plant room.
  • Accurate (3% FS), reproducible results using high quality interference filters with long-life LEDs
  • Internal ring memory for 16 data sets
  • Real time clock and date
  • Optional infrared interface for transfer of test data to PC or printer
The unit supplies accurate, reproduceable results using high quality interference filters with long-life LEDs as a light source. Tests are conducted using the standard small scale sealed tube method. Lovibond® Vario COD vials are programmed for measurements of 0-150, 0-1500 or 0-15000 mg/l.
Delivery information: Instrument supplied with carry case, 4 x AAA batteries, 3 x round glass vials with lids, plastic syringe (1 and 2 mL), COD vial adapter, guarantee sheet, certificate (COC), and instruction manual.